siRNA Details
| Gene Symbol: Slc6a6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017206.1 | NP_058902.1 | 3, 4 | 363 | ||
| XM_006236903.2 | XP_006236965.1 | 2, 3 | 472 | |||
| XM_008763123.1 | XP_008761345.1 | 4, 5 | 633 | |||
| GenBank mRNA | M96601.1 | - | 3, 4 | 363 | ||

Slc6a6
Genomic Map