siRNA Details
| Gene Symbol: Pitpna | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017231.1 | NP_058927.1 | 3 | 350 | ||
| GenBank mRNA | BC070945.1 | - | 3 | 378 | 7103390 | |
| M25758.1 | - | 3 | 350 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pitpna| Gene Symbol: Pitpna | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017231.1 | NP_058927.1 | 3 | 350 | ||
| GenBank mRNA | BC070945.1 | - | 3 | 378 | 7103390 | |
| M25758.1 | - | 3 | 350 | |||