siRNA Details
| Gene Symbol: Casq2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017131.2 | NP_058827.3 | 1, 2 | 379 | ||
| GenBank mRNA | BC072547.1 | - | 1, 2 | 366 | 7105911 | |
| CK602439.1 | - | 1, 2 | 379 | |||
| U33287.1 | - | 1, 2 | 360 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Casq2| Gene Symbol: Casq2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017131.2 | NP_058827.3 | 1, 2 | 379 | ||
| GenBank mRNA | BC072547.1 | - | 1, 2 | 366 | 7105911 | |
| CK602439.1 | - | 1, 2 | 379 | |||
| U33287.1 | - | 1, 2 | 360 | |||