siRNA Details
Gene Symbol: Rcn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017132.2 | NP_058828.2 | 5 | 805 | ||
GenBank mRNA | BC061962.1 | - | 5 | 759 | 5598390 | |
U15734.1 | - | 5 | 816 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Rcn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017132.2 | NP_058828.2 | 5 | 805 | ||
GenBank mRNA | BC061962.1 | - | 5 | 759 | 5598390 | |
U15734.1 | - | 5 | 816 |