siRNA Details
| Gene Symbol: Arg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017134.3 | NP_058830.2 | 4, 5 | 613 | ||
| GenBank mRNA | BC091158.1 | - | 4, 5 | 543 | 7368991 | |
| FM100195.1 | - | 4, 5 | 582 | |||
| FM120421.1 | - | 2, 3 | 255 | |||
| FQ209619.1 | - | 1, 2 | 123 | |||
| FQ218591.1 | - | 4, 5 | 557 | |||
| FQ219159.1 | - | 4, 5 | 494 | |||
| J02720.1 | - | 4, 5 | 494 | |||

Arg1
Genomic Map