siRNA Details
| Gene Symbol: Plcg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017168.1 | NP_058864.1 | 2, 3 | 395 | ||
| XM_008772607.1 | XP_008770829.1 | 2, 3 | 411 | |||
| GenBank mRNA | J05155.1 | - | 2, 3 | 395 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Plcg2| Gene Symbol: Plcg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017168.1 | NP_058864.1 | 2, 3 | 395 | ||
| XM_008772607.1 | XP_008770829.1 | 2, 3 | 411 | |||
| GenBank mRNA | J05155.1 | - | 2, 3 | 395 | ||