siRNA Details
| Gene Symbol: Prdx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017169.1 | NP_058865.1 | 5 | 537 | ||
| XM_006255244.1 | XP_006255306.1 | 5 | 718 | |||
| GenBank mRNA | BC058481.1 | - | 5 | 564 | 6921522 | |
| FQ211079.1 | - | 5 | 553 | |||
| FQ215547.1 | - | 5 | 589 | |||
| FQ221561.1 | - | 5 | 558 | |||
| FQ225602.1 | - | 5 | 560 | |||
| FQ227282.1 | - | 5 | 560 | |||
| FQ227580.1 | - | 5 | 551 | |||
| FQ233937.1 | - | 5 | 560 | |||
| FQ234358.1 | - | 5 | 589 | |||
| FQ234566.1 | - | 5 | 655 | |||
| U06099.1 | - | 5 | 537 | |||

Prdx2
Genomic Map