siRNA Details
| Gene Symbol: Pkn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017175.2 | NP_058871.2 | 5 | 839 | ||
| XM_006255245.2 | XP_006255307.1 | 5 | 1879 | |||
| GenBank mRNA | BC061836.1 | - | 5 | 740 | 5599989 | |
| D26180.1 | - | 5 | 790 | |||
| FQ222217.1 | - | 5 | 839 | |||
| L35634.1 | - | 5 | 682 | |||

Pkn1
Genomic Map