siRNA Details
| Gene Symbol: Chrne | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017194.1 | NP_058890.1 | 6 | 574 | ||
| GenBank mRNA | X13252.1 | - | 6 | 574 | ||
| X74836.1 | - | 6 | 574 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Chrne| Gene Symbol: Chrne | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017194.1 | NP_058890.1 | 6 | 574 | ||
| GenBank mRNA | X13252.1 | - | 6 | 574 | ||
| X74836.1 | - | 6 | 574 | |||