siRNA Details
| Gene Symbol: Aif1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017196.3 | NP_058892.1 | 3, 4 | 250 | ||
| XM_006256065.2 | XP_006256127.1 | 5, 6 | 400 | |||
| GenBank mRNA | AB000818.1 | - | 3, 4 | 248 | ||
| D82069.1 | - | 5, 6 | 405 | |||
| FM053942.1 | - | 3, 4 | 237 | |||
| FQ231499.1 | - | 3, 4 | 248 | |||
| U17919.1 | - | 3, 4 | 222 | |||

Aif1
Genomic Map