siRNA Details
| Gene Symbol: Ssr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017199.1 | NP_058895.1 | 3 | 409 | ||
| XM_006229571.2 | XP_006229633.1 | 3 | 465 | |||
| GenBank mRNA | BC058482.1 | - | 3 | 322 | 6921620 | |
| FQ221421.1 | - | 1 | 25 | |||
| FQ221423.1 | - | 1 | 25 | |||
| FQ221949.1 | - | 1 | 25 | |||
| FQ228311.1 | - | 1 | 25 | |||
| FQ228501.1 | - | 1 | 25 | |||
| Z19087.1 | - | 3 | 409 | |||

Ssr4
Genomic Map