siRNA Details
| Gene Symbol: Rgs4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017214.1 | NP_058910.1 | 2, 3 | 266 | ||
| XM_008769736.1 | XP_008767958.1 | 2, 3 | 295 | |||
| GenBank mRNA | AF117211.1 | - | 2, 3 | 266 | ||
| U27767.1 | - | 2, 3 | 220 | |||

Rgs4
Genomic Map