siRNA Details
| Gene Symbol: Slc22a6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017224.2 | NP_058920.1 | 6 | 998 | ||
| XM_006230978.2 | XP_006231040.1 | 6 | 1253 | |||
| GenBank mRNA | AB004559.1 | - | 6 | 1228 | ||
| AF008221.1 | - | 6 | 1206 | |||
| BC104692.1 | - | 6 | 998 | 7109188 | ||

Slc22a6
Genomic Map