siRNA Details
| Gene Symbol: Pydc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162938.1 | NP_001156410.1 | 4 | 1376 | ||
| GenBank mRNA | AK036671.1 | - | 4 | 1146 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pydc3| Gene Symbol: Pydc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162938.1 | NP_001156410.1 | 4 | 1376 | ||
| GenBank mRNA | AK036671.1 | - | 4 | 1146 | ||