siRNA Details
| Gene Symbol: Wfdc17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081957.1 | NP_001075426.1 | 3 | 453 | ||
| GenBank mRNA | BC038870.1 | - | 3 | 382 | ||
| BC089618.1 | - | 3 | 426 | 6766535 | ||
| FJ007372.1 | - | 3 | 494 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wfdc17| Gene Symbol: Wfdc17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081957.1 | NP_001075426.1 | 3 | 453 | ||
| GenBank mRNA | BC038870.1 | - | 3 | 382 | ||
| BC089618.1 | - | 3 | 426 | 6766535 | ||
| FJ007372.1 | - | 3 | 494 | |||