siRNA Details
| Gene Symbol: Fam129c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166213.1 | NP_001159685.1 | 3, 4 | 431 | ||
| GenBank mRNA | AK158308.1 | - | 3, 4 | 296 | ||
| Gene Symbol: Pgls | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK036059.1 | - | 926 | |||

Fam129c
Genomic Map