siRNA Details
| Gene Symbol: Fam129c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166213.1 | NP_001159685.1 | 15 | 2210 | ||
| GenBank mRNA | AK158308.1 | - | 13 | 2788 | ||
| AK163580.1 | - | 10 | 1515 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam129c| Gene Symbol: Fam129c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166213.1 | NP_001159685.1 | 15 | 2210 | ||
| GenBank mRNA | AK158308.1 | - | 13 | 2788 | ||
| AK163580.1 | - | 10 | 1515 | |||