siRNA Details
| Gene Symbol: Gm11744 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163318.1 | NP_001156790.1 | 1 | 148 | ||
| GenBank mRNA | AK137302.1 | - | 579 | |||
| CK620045.1 | - | 1 | 94 | |||
| DQ390337.1 | - | 1 | 148 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm11744| Gene Symbol: Gm11744 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163318.1 | NP_001156790.1 | 1 | 148 | ||
| GenBank mRNA | AK137302.1 | - | 579 | |||
| CK620045.1 | - | 1 | 94 | |||
| DQ390337.1 | - | 1 | 148 | |||