siRNA Details
| Gene Symbol: 7420461P10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177581.1 | NP_001171052.1 | 2 | 286 | ||
| GenBank mRNA | AK136134.1 | - | 2 | 241 | ||
| BC099523.1 | - | 2 | 286 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

7420461P10Rik| Gene Symbol: 7420461P10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177581.1 | NP_001171052.1 | 2 | 286 | ||
| GenBank mRNA | AK136134.1 | - | 2 | 241 | ||
| BC099523.1 | - | 2 | 286 | |||