siRNA Details
Gene Symbol: Cep85l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204983.1 | NP_001191912.1 | 12, 13 | 2523 |

More Information
Other Species Targeted

Gene Ontology Categories:

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cep85l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204983.1 | NP_001191912.1 | 12, 13 | 2523 |