siRNA Details
| Gene Symbol: Gm10406 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164727.1 | NP_001158199.1 | 7 | 1889 | ||
| Gene Symbol: Gm3696 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC055874.1 | - | 1887 | 3491523 | ||

Gm10406
Genomic Map