siRNA Details
| Gene Symbol: Ccdc152 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166063.2 | NP_001159535.1 | 6 | 419 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc152| Gene Symbol: Ccdc152 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166063.2 | NP_001159535.1 | 6 | 419 | ||