siRNA Details
| Gene Symbol: Gm6182 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC080301.1 | - | 4212 | |||
| BC090643.1 | - | 3944 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm6182| Gene Symbol: Gm6182 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC080301.1 | - | 4212 | |||
| BC090643.1 | - | 3944 | ||||