siRNA Details
| Gene Symbol: Gm6182 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK008984.1 | - | 6 | 670 | ||
| AK166086.1 | - | 5 | 573 | |||
| BC048160.1 | - | 5 | 536 | |||
| BC051922.1 | - | 5 | 536 | |||
| BC059210.1 | - | 4 | 1670 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm6182| Gene Symbol: Gm6182 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK008984.1 | - | 6 | 670 | ||
| AK166086.1 | - | 5 | 573 | |||
| BC048160.1 | - | 5 | 536 | |||
| BC051922.1 | - | 5 | 536 | |||
| BC059210.1 | - | 4 | 1670 | |||