siRNA Details
| Gene Symbol: Gm14920 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001102665.1 | NP_001096135.1 | 1 | 253 | ||
| Gene Symbol: H2afb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001281531.1 | NP_001268460.1 | 1 | 253 | ||

Gm14920
Genomic Map