siRNA Details
| Gene Symbol: Gm12942 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099319.2 | NP_001092789.1 | 3 | 788 | ||
| GenBank mRNA | AK034325.1 | - | 1 | 3271 | ||
| AK156456.1 | - | 3 | 770 | |||
| AK160613.1 | - | 1 | 3265 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm12942| Gene Symbol: Gm12942 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099319.2 | NP_001092789.1 | 3 | 788 | ||
| GenBank mRNA | AK034325.1 | - | 1 | 3271 | ||
| AK156456.1 | - | 3 | 770 | |||
| AK160613.1 | - | 1 | 3265 | |||