siRNA Details
| Gene Symbol: Polb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017141.2 | NP_058837.2 | 8 | 452 | ||
| GenBank mRNA | BC098668.1 | - | 8 | 452 | 7386894 | |
| J02776.1 | - | 8 | 440 | |||
| M13961.1 | - | 8 | 430 | |||
| U38801.1 | - | 447 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Polb| Gene Symbol: Polb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017141.2 | NP_058837.2 | 8 | 452 | ||
| GenBank mRNA | BC098668.1 | - | 8 | 452 | 7386894 | |
| J02776.1 | - | 8 | 440 | |||
| M13961.1 | - | 8 | 430 | |||
| U38801.1 | - | 447 | ||||