siRNA Details
| Gene Symbol: Cyp4a32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100181.1 | NP_001093651.1 | 13 | 1767 | ||
| Gene Symbol: Cyp4a10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | X69296.1 | - | 1740 | |||

Cyp4a32
Genomic Map