siRNA Details
| Gene Symbol: G630090E17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001173500.2 | NP_001166971.1 | 2 | 341 | ||
| GenBank mRNA | AK144527.1 | - | 2 | 342 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

G630090E17Rik| Gene Symbol: G630090E17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001173500.2 | NP_001166971.1 | 2 | 341 | ||
| GenBank mRNA | AK144527.1 | - | 2 | 342 | ||