siRNA Details
| Gene Symbol: Gm16401 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK141347.1 | - | 1 | 1720 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm16401| Gene Symbol: Gm16401 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK141347.1 | - | 1 | 1720 | ||