siRNA Details
| Gene Symbol: Gm13247 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243138.1 | NP_001230067.1 | 5 | 1906 | ||
| NM_001243139.1 | NP_001230068.1 | 4 | 1791 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm13247| Gene Symbol: Gm13247 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243138.1 | NP_001230067.1 | 5 | 1906 | ||
| NM_001243139.1 | NP_001230068.1 | 4 | 1791 | |||