siRNA Details
| Gene Symbol: Cyp3a59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105160.1 | NP_001098630.1 | 13 | 1683 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cyp3a59| Gene Symbol: Cyp3a59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105160.1 | NP_001098630.1 | 13 | 1683 | ||