siRNA Details
| Gene Symbol: Gm20877 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199332.1 | NP_001186261.1 | 2 | 230 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm20877| Gene Symbol: Gm20877 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199332.1 | NP_001186261.1 | 2 | 230 | ||