siRNA Details
| Gene Symbol: 9030025P20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001123370.1 | NP_001116842.1 | 10 | 1002 | ||
| GenBank mRNA | AK042420.1 | - | 9 | 1935 | ||
| AK078855.1 | - | 10 | 990 | |||
| AK160486.1 | - | 10 | 1054 | |||
| AK161147.1 | - | 9 | 1797 | |||

9030025P20Rik
Genomic Map