siRNA Details
| Gene Symbol: 9030025P20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001123370.1 | NP_001116842.1 | 12 | 1625 | ||
| GenBank mRNA | AK161147.1 | - | 11 | 2420 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

9030025P20Rik| Gene Symbol: 9030025P20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001123370.1 | NP_001116842.1 | 12 | 1625 | ||
| GenBank mRNA | AK161147.1 | - | 11 | 2420 | ||