siRNA Details
Gene Symbol: Gm3604 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162910.1 | NP_001156382.1 | 4 | 1529 | ||
GenBank mRNA | BC049144.1 | - | 4 | 1521 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm3604 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162910.1 | NP_001156382.1 | 4 | 1529 | ||
GenBank mRNA | BC049144.1 | - | 4 | 1521 |