siRNA Details
| Gene Symbol: Gm3696 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024712.2 | NP_001019883.2 | 774 | |||
| GenBank mRNA | EF651802.1 | - | 405 | |||
| EF651822.1 | - | 3 | 405 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm3696| Gene Symbol: Gm3696 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024712.2 | NP_001019883.2 | 774 | |||
| GenBank mRNA | EF651802.1 | - | 405 | |||
| EF651822.1 | - | 3 | 405 | |||