siRNA Details
| Gene Symbol: Gm10639 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122660.1 | NP_001116132.1 | 5 | 448 | ||
| GenBank mRNA | BC030173.1 | - | 5 | 459 | 5041881 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm10639| Gene Symbol: Gm10639 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122660.1 | NP_001116132.1 | 5 | 448 | ||
| GenBank mRNA | BC030173.1 | - | 5 | 459 | 5041881 | |