siRNA Details
Gene Symbol: Cyp2b12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017156.1 | NP_058852.1 | 767 | |||
GenBank mRNA | X63545.1 | - | 767 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cyp2b12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017156.1 | NP_058852.1 | 767 | |||
GenBank mRNA | X63545.1 | - | 767 |