siRNA Details
Gene Symbol: Vmn2r55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001104645.1 | NP_001098115.1 | 2 | 578 | ||
GenBank mRNA | AB919108.1 | - | 2 | 578 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
