siRNA Details
| Gene Symbol: Vmn2r45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105075.1 | NP_001098545.1 | 3 | 1164 | ||
 Back To Top
 Back To TopMore Information
| Set Membership: |  G-protein coupled receptor | 
Panther Classification:
Gene Ontology Categories:
 Back To Top
 Back To Top
 Vmn2r45
Vmn2r45 Genomic Map
Genomic Map Back To Top
 Back To Top