siRNA Details
| Gene Symbol: Scgb2b3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270541.1 | NP_001257470.1 | 3 | 300 | ||
| GenBank mRNA | GU269235.1 | - | 3 | 300 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Scgb2b3| Gene Symbol: Scgb2b3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270541.1 | NP_001257470.1 | 3 | 300 | ||
| GenBank mRNA | GU269235.1 | - | 3 | 300 | ||