siRNA Details
| Gene Symbol: Slc6a18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017163.1 | NP_058859.1 | 6, 7 | 880 | ||
| XM_006227772.2 | XP_006227834.1 | 6, 7 | 968 | |||
| XM_006227774.2 | XP_006227836.1 | 6, 7 | 968 | |||
| XM_006227775.2 | XP_006227837.1 | 6, 7 | 968 | |||
| XM_008758682.1 | XP_008756904.1 | 5, 6 | 857 | |||
| GenBank mRNA | U12973.1 | - | 6, 7 | 880 | ||

Slc6a18
Genomic Map