siRNA Details
| Gene Symbol: Scgb2b7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001198871.1 | NP_001185800.1 | 2 | 92 | ||
| GenBank mRNA | CK615801.1 | - | 2 | 93 | ||
| CK616838.1 | - | 2 | 89 | |||
| CK616903.1 | - | 2 | 90 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Scgb2b7| Gene Symbol: Scgb2b7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001198871.1 | NP_001185800.1 | 2 | 92 | ||
| GenBank mRNA | CK615801.1 | - | 2 | 93 | ||
| CK616838.1 | - | 2 | 89 | |||
| CK616903.1 | - | 2 | 90 | |||