siRNA Details
Gene Symbol: Klrb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099918.1 | NP_001093388.1 | 4, 5 | 402 | ||
GenBank mRNA | DQ113419.1 | - | 4, 5 | 402 | ||
HQ713436.1 | - | 4, 5 | 402 | |||
JF429832.1 | - | 4, 5 | 415 |
