siRNA Details
| Gene Symbol: R3hdml | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099331.2 | NP_001092801.1 | 2 | 511 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

R3hdml| Gene Symbol: R3hdml | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099331.2 | NP_001092801.1 | 2 | 511 | ||