siRNA Details
| Gene Symbol: Rnf223 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001220499.1 | NP_001207428.1 | 1 | 767 | ||
| GenBank mRNA | AK137636.1 | - | 1 | 2424 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rnf223| Gene Symbol: Rnf223 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001220499.1 | NP_001207428.1 | 1 | 767 | ||
| GenBank mRNA | AK137636.1 | - | 1 | 2424 | ||