siRNA Details
| Gene Symbol: Camta1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081557.3 | NP_001075026.1 | 21 | 4827 | ||
| GenBank mRNA | AK122383.1 | - | 16 | 4253 | ||
| AK132560.1 | - | 8 | 1610 | |||
| AK147482.1 | - | 11 | 1634 | |||
| AK168378.1 | - | 10 | 1539 | |||
| BC150740.1 | - | 20 | 4337 | 9087651 | ||

Camta1
Genomic Map