siRNA Details
| Gene Symbol: 5430421N21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001201323.1 | NP_001188252.1 | 4 | 751 | ||
| GenBank mRNA | AK133718.1 | - | 4 | 752 | ||
| M92088.1 | - | 4 | 736 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

5430421N21Rik| Gene Symbol: 5430421N21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001201323.1 | NP_001188252.1 | 4 | 751 | ||
| GenBank mRNA | AK133718.1 | - | 4 | 752 | ||
| M92088.1 | - | 4 | 736 | |||