siRNA Details
Gene Symbol: Esp31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177586.1 | NP_001171057.1 | 3 | 777 | ||
GenBank mRNA | AB307010.1 | - | 3 | 777 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Esp31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177586.1 | NP_001171057.1 | 3 | 777 | ||
GenBank mRNA | AB307010.1 | - | 3 | 777 |